Methyl 1-(2-Methoxyphenyl)-4-oxocyclohexanecarboxylate

Lieferant: Apollo Scientific

OR470528-5G
APOSOR470528-5GEA 1010 EUR
APOSOR470528-5G
Methyl 1-(2-Methoxyphenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(2-Methoxyphenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₅H₁₈O₄
Molecular Weight: 262,301 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423129
CAS-Nummer: 1385694-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR