Methyl 1-(2-Nitrophenyl)-4-oxocyclohexanecarboxylate

Lieferant: Apollo Scientific

OR470531-5G
APOSOR470531-5GEA 1010 EUR
APOSOR470531-5G
Methyl 1-(2-Nitrophenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(2-Nitrophenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₄H₁₅NO₅
Molecular Weight: 277,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423137
CAS-Nummer: 1385694-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR