1-(2-Chlorophenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

OR470532-5G
APOSOR470532-5GEA 1010 EUR
APOSOR470532-5G
1-(2-Chlorophenyl)-4-oxocyclohexanecarboxylic acid
1-(2-Chlorophenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₃H₁₃ClO₃
Molecular Weight: 252,694 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423138
CAS-Nummer: 1385694-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR