Methyl 1-(2-Chlorophenyl)-4-oxocyclohexanecarboxylate
Lieferant: Apollo Scientific
OR470533-5G
APOSOR470533-5GEA
1010
EUR
APOSOR470533-5G
Methyl 1-(2-Chlorophenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(2-Chlorophenyl)-4-oxocyclohexanecarboxylate
Formel:
C₁₄H₁₅ClO₃ Molecular Weight: 266,72 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22423139 CAS-Nummer: 1385694-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...