Methyl 4-Oxo-1-(o-tolyl)cyclohexanecarboxylate

Lieferant: Apollo Scientific

OR470535-5G
APOSOR470535-5GEA 1010 EUR
APOSOR470535-5G
Methyl 4-Oxo-1-(o-tolyl)cyclohexanecarboxylate
Methyl 4-Oxo-1-(o-tolyl)cyclohexanecarboxylate
Formel: C₁₅H₁₈O₃
Molecular Weight: 246,302 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423142
CAS-Nummer: 1385694-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR