1-(2-Methyl-3-nitrophenyl)-4-oxocyclohexanecarboxylic acid
Lieferant: Apollo Scientific
OR470536-5G
APOSOR470536-5GEA
1120
EUR
APOSOR470536-5G
1-(2-Methyl-3-nitrophenyl)-4-oxocyclohexanecarboxylic acid
1-(2-Methyl-3-nitrophenyl)-4-oxocyclohexanecarboxylic acid
Formel:
C₁₄H₁₅NO₅ Molecular Weight: 277,273 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22423143 CAS-Nummer: 1385694-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...