1-(3-Bromophenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

OR470538-5G
APOSOR470538-5GEA 1010 EUR
APOSOR470538-5G
1-(3-Bromophenyl)-4-oxocyclohexanecarboxylic acid
1-(3-Bromophenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₃H₁₃BrO₃
Molecular Weight: 297,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423145
CAS-Nummer: 1385694-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR