Methyl 1-(3,4-Dichlorophenyl)-4-oxocyclohexanecarboxylate

Lieferant: Apollo Scientific

OR470539-5G
APOSOR470539-5GEA 1010 EUR
APOSOR470539-5G
Methyl 1-(3,4-Dichlorophenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(3,4-Dichlorophenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₄H₁₄Cl₂O₃
Molecular Weight: 301,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423147
CAS-Nummer: 1384264-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR