1-(2-Bromophenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

OR470540-5G
APOSOR470540-5GEA 1010 EUR
APOSOR470540-5G
1-(2-Bromophenyl)-4-oxocyclohexanecarboxylic acid
1-(2-Bromophenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₃H₁₃BrO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22423148
CAS-Nummer: 1385694-47-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR