Methyl 1-(2-Bromophenyl)-4-oxocyclohexanecarboxylate

Lieferant: Apollo Scientific

OR470541-5G
APOSOR470541-5GEA 1010 EUR
APOSOR470541-5G
Methyl 1-(2-Bromophenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(2-Bromophenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₄H₁₅BrO₃
Molecular Weight: 311,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423149
CAS-Nummer: 1385694-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR