Ethyl 1-(4-Nitrophenyl)-4-oxocyclohexanecarboxylate

Lieferant: Apollo Scientific

OR470542-5G
APOSOR470542-5GEA 1010 EUR
APOSOR470542-5G
Ethyl 1-(4-Nitrophenyl)-4-oxocyclohexanecarboxylate
Ethyl 1-(4-Nitrophenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₅H₁₇NO₅
Molecular Weight: 291,299 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22574980
CAS-Nummer: 1408058-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR