2-Amino-4-(aminomethyl)pyridine dihydrochloride

Lieferant: Apollo Scientific

OR470543-1G
APOSOR470543-1GEA 601 EUR
APOSOR470543-1G
2-Amino-4-(aminomethyl)pyridine dihydrochloride
2-Amino-4-(aminomethyl)pyridine dihydrochloride
Formel: C₆H₉N₃·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD20040454
CAS-Nummer: 618446-34-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR