Ethyl 1-(2-Chlorophenyl)-4-oxocyclohexanecarboxylate

Lieferant: Apollo Scientific

OR470544-5G
APOSOR470544-5GEA 1010 EUR
APOSOR470544-5G
Ethyl 1-(2-Chlorophenyl)-4-oxocyclohexanecarboxylate
Ethyl 1-(2-Chlorophenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₅H₁₇ClO₃
Molecular Weight: 280,747 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423150
CAS-Nummer: 1385694-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR