N-Cbz-4-(1,3-dioxolan-2-yl)aniline

Lieferant: Apollo Scientific

OR470545-5G
APOSOR470545-5GEA 397 EUR
APOSOR470545-5G
N-Cbz-4-(1,3-dioxolan-2-yl)aniline
N-Cbz-4-(1,3-dioxolan-2-yl)aniline
Formel: C₁₇H₁₇NO₄
Molecular Weight: 299,321 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22375700
CAS-Nummer: 1224886-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR