(3-Pyridyl)(3-quinolyl)methanol
Lieferant: Apollo Scientific
OR470549-1G
APOSOR470549-1GEA
499
EUR
APOSOR470549-1G
(3-Pyridyl)(3-quinolyl)methanol
(3-Pyridyl)(3-quinolyl)methanol
Formel:
C₁₅H₁₂N₂O Molecular Weight: 236,269 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12833812 CAS-Nummer: 1183608-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...