1-Phenyl-1-(3-quinolyl)ethanol
Lieferant: Apollo Scientific
OR470550-1G
APOSOR470550-1GEA
601
EUR
APOSOR470550-1G
1-Phenyl-1-(3-quinolyl)ethanol
1-Phenyl-1-(3-quinolyl)ethanol
Formel:
C₁₇H₁₅NO Molecular Weight: 249,307 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12833780 CAS-Nummer: 1183075-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...