2-(4-Benzyl-2-morpholinyl)ethanamine

Lieferant: Apollo Scientific

OR470551-1G
APOSOR470551-1GEA 916 EUR
APOSOR470551-1G
2-(4-Benzyl-2-morpholinyl)ethanamine
2-(4-Benzyl-2-morpholinyl)ethanamine
Formel: C₁₃H₂₀N₂O
Molecular Weight: 220,311 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11108389
CAS-Nummer: 112914-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR