(4-Bromo-2-chlorophenyl)(4-chlorophenyl)methanone

Lieferant: Apollo Scientific

OR470552-5G
APOSOR470552-5GEA 1120 EUR
APOSOR470552-5G
(4-Bromo-2-chlorophenyl)(4-chlorophenyl)methanone
(4-Bromo-2-chlorophenyl)(4-chlorophenyl)methanone
Formel: C₁₃H₇BrCl₂O
Molecular Weight: 330,004 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12065414
CAS-Nummer: 1157395-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR