8-Methyl-3,4-dihydroquinolin-2(1H)-one

Lieferant: Apollo Scientific

OR470553-5G
APOSOR470553-5GEA 997 EUR
APOSOR470553-5G
8-Methyl-3,4-dihydroquinolin-2(1H)-one
8-Methyl-3,4-dihydroquinolin-2(1H)-one
Formel: C₁₀H₁₁NO
Molecular Weight: 161,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00464327
CAS-Nummer: 20151-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR