Ethyl 10-Bromo-9-anthracenecarboxylate
Lieferant: Apollo Scientific
Synonyme:
Ethyl 10-bromoanthracene-9-carboxylate
OR470554-1G
APOSOR470554-1GEA
114
EUR
APOSOR470554-1G
Ethyl 10-Bromo-9-anthracenecarboxylate
Ethyl 10-Bromo-9-anthracenecarboxylate
Formel:
C₁₇H₁₃BrO₂ Molecular Weight: 329.188 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22571679 CAS-Nummer: 1089318-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...