2-(Cbz-amino)propanamide

Lieferant: Apollo Scientific

OR470555-5G
APOSOR470555-5GEA 499 EUR
APOSOR470555-5G
2-(Cbz-amino)propanamide
2-(Cbz-amino)propanamide
Formel: C₁₁H₁₄N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00136699
CAS-Nummer: 2503-29-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR