7-Hydroxypyrazolo[1,5-a]pyrimidine-6-carbonitrile

Lieferant: Apollo Scientific

OR470558-1G
APOSOR470558-1GEA 448 EUR
APOSOR470558-1G
7-Hydroxypyrazolo[1,5-a]pyrimidine-6-carbonitrile
7-Hydroxypyrazolo[1,5-a]pyrimidine-6-carbonitrile
Formel: C₇H₄N₄O
Molecular Weight: 160,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00973929
CAS-Nummer: 89642-01-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR