3-Bromo-7-chlorothieno[3,2-b]pyridine-6-carbonitrile

Lieferant: Apollo Scientific

OR470559-1G
APOSOR470559-1GEA 713 EUR
APOSOR470559-1G
3-Bromo-7-chlorothieno[3,2-b]pyridine-6-carbonitrile
3-Bromo-7-chlorothieno[3,2-b]pyridine-6-carbonitrile
Formel: C₈H₂BrClN₂S
Molecular Weight: 273,537 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14706347
CAS-Nummer: 798574-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR