3-Amino-2-indenecarbonitrile

Lieferant: Apollo Scientific

OR470560-1G
APOSOR470560-1GEA 448 EUR
APOSOR470560-1G
3-Amino-2-indenecarbonitrile
3-Amino-2-indenecarbonitrile
Formel: C₁₀H₈N₂
Molecular Weight: 156.184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01104042
CAS-Nummer: 195067-12-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR