4-(3-Pyridyl)-3-butyn-1-ol

Lieferant: Apollo Scientific

Synonyme: 4-(Pyridin-3-yl)but-3-yn-1-ol

OR470563-5G
APOSOR470563-5GEA 331 EUR
APOSOR470563-5G
4-(3-Pyridyl)-3-butyn-1-ol
4-(3-Pyridyl)-3-butyn-1-ol
Formel: C₉H₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD12808175
CAS-Nummer: 138487-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR