3-(4-Pyridyl)-2-propen-1-ol

Lieferant: Apollo Scientific

OR470566-5G
APOSOR470566-5GEA 916 EUR
APOSOR470566-5G
3-(4-Pyridyl)-2-propen-1-ol
3-(4-Pyridyl)-2-propen-1-ol
Formel: C₈H₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD18384354
CAS-Nummer: 193002-34-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR