Methyl 4-(Trimethylsilyl)benzoate

Lieferant: Apollo Scientific

OR470568-5G
APOSOR470568-5GEA 205 EUR
APOSOR470568-5G
Methyl 4-(Trimethylsilyl)benzoate
Methyl 4-(Trimethylsilyl)benzoate
Formel: C₁₁H₁₆O₂Si
Lagertemperatur: Raumtemperatur
MDL: MFCD22682799
CAS-Nummer: 22515-30-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR