4-(Trimethylsilyl)phenyl Pivalate
Lieferant: Apollo Scientific
OR470570-5G
APOSOR470570-5GEA
306
EUR
APOSOR470570-5G
4-(Trimethylsilyl)phenyl Pivalate
4-(Trimethylsilyl)phenyl Pivalate
Formel:
C₁₄H₂₂O₂Si Lagertemperatur: Raumtemperatur |
MDL:
MFCD22682787 CAS-Nummer: 1418117-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...