4-(2-Hydroxyethyl)-4H-1,2,4-triazole

Lieferant: Apollo Scientific

Synonyme: 2-(4H-1,2,4-Triazol-4-yl)ethanol

OR470572-5G
APOSOR470572-5GEA 671 EUR
APOSOR470572-5G
4-(2-Hydroxyethyl)-4H-1,2,4-triazole
4-(2-Hydroxyethyl)-4H-1,2,4-triazole
Formel: C₄H₇N₃O
Molecular Weight: 113.118 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03265455
CAS-Nummer: 66760-19-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR