Methyl 3-(4-Methyl-2,5-dioxo-4-imidazolidinyl)propanoate

Lieferant: Apollo Scientific

OR470573-1G
APOSOR470573-1GEA 397 EUR
APOSOR470573-1G
Methyl 3-(4-Methyl-2,5-dioxo-4-imidazolidinyl)propanoate
Methyl 3-(4-Methyl-2,5-dioxo-4-imidazolidinyl)propanoate
Formel: C₈H₁₂N₂O₄
Molecular Weight: 200,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22682789
CAS-Nummer: 1418117-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR