Ethyl 3-(4-Methyl-2,5-dioxo-4-imidazolidinyl)propanoate
Lieferant: Apollo Scientific
OR470574-1G
APOSOR470574-1GEA
397
EUR
APOSOR470574-1G
Ethyl 3-(4-Methyl-2,5-dioxo-4-imidazolidinyl)propanoate
Ethyl 3-(4-Methyl-2,5-dioxo-4-imidazolidinyl)propanoate
Formel:
C₉H₁₄N₂O₄ Molecular Weight: 214,218 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22682790 CAS-Nummer: 1418117-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...