Methyl 2-[(5-Bromo-2-pyridyl)oxy]acetate

Lieferant: Apollo Scientific

OR470575-5G
APOSOR470575-5GEA 713 EUR
APOSOR470575-5G
Methyl 2-[(5-Bromo-2-pyridyl)oxy]acetate
Methyl 2-[(5-Bromo-2-pyridyl)oxy]acetate
Formel: C₈H₈BrNO₃
Molecular Weight: 246,058 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13190760
CAS-Nummer: 845890-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR