Ethyl 5-Methoxy-2-thiophenecarboxylate
Lieferant: Apollo Scientific
OR470577-1G
APOSOR470577-1GEA
601
EUR
APOSOR470577-1G
Ethyl 5-Methoxy-2-thiophenecarboxylate
Ethyl 5-Methoxy-2-thiophenecarboxylate
Formel:
C₈H₁₀O₃S Molecular Weight: 186,228 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22682791 CAS-Nummer: 1418117-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...