5-Ethoxy-2-thiophenecarboxylic acid

Lieferant: Apollo Scientific

OR470578-1G
APOSOR470578-1GEA 601 EUR
APOSOR470578-1G
5-Ethoxy-2-thiophenecarboxylic acid
5-Ethoxy-2-thiophenecarboxylic acid
Formel: C₇H₈O₃S
Molecular Weight: 172,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08692953
CAS-Nummer: 135080-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR