Methyl 5-Ethoxy-2-thiophenecarboxylate

Lieferant: Apollo Scientific

OR470579-1G
APOSOR470579-1GEA 601 EUR
APOSOR470579-1G
Methyl 5-Ethoxy-2-thiophenecarboxylate
Methyl 5-Ethoxy-2-thiophenecarboxylate
Formel: C₈H₁₀O₃S
Molecular Weight: 186,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22682792
CAS-Nummer: 1418117-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR