Ethyl 5-Ethoxy-2-thiophenecarboxylate

Lieferant: Apollo Scientific

OR470580-1G
APOSOR470580-1GEA 601 EUR
APOSOR470580-1G
Ethyl 5-Ethoxy-2-thiophenecarboxylate
Ethyl 5-Ethoxy-2-thiophenecarboxylate
Formel: C₉H₁₂O₃S
Molecular Weight: 200,255 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22682793
CAS-Nummer: 1418117-85-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR