4-(3-Aminopropyl)-4H-1,2,4-triazole

Lieferant: Apollo Scientific

OR470581-1G
APOSOR470581-1GEA 601 EUR
APOSOR470581-1G
4-(3-Aminopropyl)-4H-1,2,4-triazole
4-(3-Aminopropyl)-4H-1,2,4-triazole
Formel: C₅H₁₀N₄
Molecular Weight: 126.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03093040
CAS-Nummer: 897054-15-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR