Ethyl 3-(4-Methyl-1-piperazinyl)propanoate

Lieferant: Apollo Scientific

OR470586-5G
APOSOR470586-5GEA 374 EUR
APOSOR470586-5G
Ethyl 3-(4-Methyl-1-piperazinyl)propanoate
Ethyl 3-(4-Methyl-1-piperazinyl)propanoate
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00785764
CAS-Nummer: 7148-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR