Ethyl 4-(4-Methyl-1-piperazinyl)butanoate

Lieferant: Apollo Scientific

OR470587-5G
APOSOR470587-5GEA 1010 EUR
APOSOR470587-5G
Ethyl 4-(4-Methyl-1-piperazinyl)butanoate
Ethyl 4-(4-Methyl-1-piperazinyl)butanoate
Formel: C₁₁H₂₂N₂O₂
Molecular Weight: 214,305 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01876880
CAS-Nummer: 487008-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR