3-(Cyclopentyloxy)-4-methoxyphenylacetonitrile

Lieferant: Apollo Scientific

OR470588-5G
APOSOR470588-5GEA 1350 EUR
APOSOR470588-5G
3-(Cyclopentyloxy)-4-methoxyphenylacetonitrile
3-(Cyclopentyloxy)-4-methoxyphenylacetonitrile
Formel: C₁₄H₁₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09834353
CAS-Nummer: 141333-36-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR