Ethyl 2-(4-Acetylphenyl)acetate
Lieferant: Apollo Scientific
OR470590-1G
APOSOR470590-1GEA
713
EUR
APOSOR470590-1G
Ethyl 2-(4-Acetylphenyl)acetate
Ethyl 2-(4-Acetylphenyl)acetate
Formel:
C₁₂H₁₄O₃ Molecular Weight: 206,238 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10698673 CAS-Nummer: 1528-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...