2-Propoxy-phenylamine

Lieferant: Apollo Scientific

Synonyme: 2-Propoxyaniline

OR470591-5G
APOSOR470591-5GEA 916 EUR
APOSOR470591-5G
2-Propoxy-phenylamine
2-Propoxy-phenylamine
Formel: C₉H₁₃NO
Molecular Weight: 151,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00628992
CAS-Nummer: 4469-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR