1-(Benzyloxy)-4-methylpyrrolidine-2,3-dione

Lieferant: Apollo Scientific

OR470595-5G
APOSOR470595-5GEA 1410 EUR
APOSOR470595-5G
1-(Benzyloxy)-4-methylpyrrolidine-2,3-dione
1-(Benzyloxy)-4-methylpyrrolidine-2,3-dione
Formel: C₁₂H₁₃NO₃
Molecular Weight: 219,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23381119
CAS-Nummer: 130865-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR