Ethyl 3-Methyl-2-oxo-3-butenoate

Lieferant: Apollo Scientific

OR470596-1G
APOSOR470596-1GEA 804 EUR
APOSOR470596-1G
Ethyl 3-Methyl-2-oxo-3-butenoate
Ethyl 3-Methyl-2-oxo-3-butenoate
Formel: C₇H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD23381118
CAS-Nummer: 50331-71-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR