3-(2-Piperidyl)propyl Acetate Acetate

Lieferant: Apollo Scientific

OR470597-5G
APOSOR470597-5GEA 1010 EUR
APOSOR470597-5G
3-(2-Piperidyl)propyl Acetate Acetate
3-(2-Piperidyl)propyl Acetate Acetate
Formel: C₁₀H₁₉NO₂·CH₃CO₂H
Molecular Weight: 245,315 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23381117
CAS-Nummer: 1427475-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR