3-(4-Piperidyl)propyl Acetate Acetate

Lieferant: Apollo Scientific

OR470599-5G
APOSOR470599-5GEA 1010 EUR
APOSOR470599-5G
3-(4-Piperidyl)propyl Acetate Acetate
3-(4-Piperidyl)propyl Acetate Acetate
Formel: C₁₀H₁₉NO₂·CH₃CO₂H
Molecular Weight: 245,315 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23381115
CAS-Nummer: 1427475-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR