Decahydroquinoxaline
Lieferant: Apollo Scientific
OR470600-1G
APOSOR470600-1GEA
916
EUR
APOSOR470600-1G
Decahydroquinoxaline
Decahydroquinoxaline
Formel:
C₈H₁₆N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092162 CAS-Nummer: 90410-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...