3-(Aminomethyl)indoline

Lieferant: Apollo Scientific

OR470603-1G
APOSOR470603-1GEA 804 EUR
APOSOR470603-1G
3-(Aminomethyl)indoline
3-(Aminomethyl)indoline
Formel: C₉H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19215168
CAS-Nummer: 933698-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR