N-(3-Indolylmethyl)benzylamine

Lieferant: Apollo Scientific

OR470605-1G OR470605-5G
APOSOR470605-1GEA 306 EUR
APOSOR470605-1G APOSOR470605-5G
N-(3-Indolylmethyl)benzylamine
N-(3-Indolylmethyl)benzylamine
Formel: C₁₆H₁₆N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD17430949
CAS-Nummer: 57506-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR