Ethyl 3-(3-Pyridyl)propanoate
Lieferant: Apollo Scientific
OR470607-5G
APOSOR470607-5GEA
713
EUR
APOSOR470607-5G
Ethyl 3-(3-Pyridyl)propanoate
Ethyl 3-(3-Pyridyl)propanoate
Formel:
C₁₀H₁₃NO₂ Molecular Weight: 179,216 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11045824 CAS-Nummer: 64107-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...